Perchloric acid
Synonym(s):
PCA, Perchloric acid, Perchloric acid solution
Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
Products
Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Reagent Type
Special Grade
Technique
Showing 1-3 of 3