Skip to Content
Merck
Sodium propionate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sodium propionate

Sodium propionate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Propionic acid sodium salt
Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique