Skip to Content
Merck
Ascentis® Phenyl HPLC Column 5 μm particle size, L × I.D. 5 cm × 4.6 mm

Ascentis® Phenyl HPLC Column

Ascentis® Phenyl HPLC Column 5 μm particle size, L × I.D. 5 cm × 4.6 mm

Products

Shipping
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique