Skip to Content
Merck
Streptomycin sulfate salt powder, BioReagent, suitable for cell culture

Streptomycin sulfate salt

Streptomycin sulfate salt powder, BioReagent, suitable for cell culture
Synonym(s):
Streptomycin Sulfate, Streptomyces sp.
Linear Formula:
C21H39N7O12 · 1.5 H2SO4
CAS Number:
Molecular Weight:
728.69

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Technique