Skip to Content
Merck
Insulin from bovine pancreas powder, BioReagent, suitable for cell culture

Insulin from bovine pancreas

Insulin from bovine pancreas powder, BioReagent, suitable for cell culture
Synonym(s):
Insulin from bovine pancreas
Empirical Formula (Hill Notation):
C254H377N65O75S6
CAS Number:
Molecular Weight:
5733.49

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Sterilization Method
Technique