Adenosine 5′-triphosphate disodium salt hydrate
Synonym(s):
ATP disodium salt, 5′-ATP-Na2, ATP disodium salt hydrate, ATP disodium hydrate
Linear Formula:
C10H14N5O13P3Na2 · xH2O
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Package Size
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing