Paroxetine hydrochloride hemihydrate
Synonym(s):
(3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate
Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · .5 H2O
CAS Number:
Molecular Weight:
374.83
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing