Skip to Content
Merck
Acetic anhydride

Acetic anhydride

Acetic anhydride
Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technique