Skip to Content
KR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Perchloric acid
Synonym(s):
PCA, Perchloric acid
Linear Formula:
HClO
4
CAS Number:
7601-90-3
Molecular Weight:
100.46
Products
Shipping
Ships Today (2)
Product Category
inorganic acids (2)
Brand
Sigma-Aldrich (3)
Supelco (2)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
244252
Perchloric acid,
ACS reagent, 70%
Expand
View Pricing
1.00519
Perchloric acid,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
311421
Perchloric acid,
70%, 99.999% trace metals basis
Expand
View Pricing
30755
Perchloric acid,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Expand
View Pricing
1.00514
Perchloric acid,
for analysis (max. 0.0000005% Hg) EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing