GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP®-2560
![GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP®-2560 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/238/732/cadd7f6b-f50f-4c58-bbdf-77ce23ceda71/800/cadd7f6b-f50f-4c58-bbdf-77ce23ceda71.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SP-2560, 100 m x 0.25 mm I.D., 0.20 μm (24056) (24056)
oven
175 °C
inj. temp.
210 °C
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec @ 175 °C
injection
1.0 μL, 100:1 split
sample
Mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC