MilliporeSigma
A Therapeutic Peptide Vaccine Against PCSK9.
Scientific reports (2017-10-04)
Yajie Pan, Yanzhao Zhou, Hailang Wu, Xiao Chen, Xiajun Hu, Hongrong Zhang, Zihua Zhou, Zhihua Qiu, Yuhua Liao
ABSTRACT
Vaccination provides a promising approach for treatment of hypercholesterolemia and improvement in compliance. In this study, the appropriate virus-like particle (VLP)-peptide vaccines targeting proprotein convertase subtilisin/kexin type 9 (PCSK9) were screened. The screening criteria of target peptides were as follows: (1) located in catalytic domain of PCSK9, or regulating the binding of PCSK9 and LDL receptors (LDLR); (2) having low/no-similarity when matched with the host proteome; (3) possessing ideal antigenicity and hydrophilicity; (4) including the functional mutation site of PCSK9. It was found that mice vaccinated with VLP -PCSK9 peptide vaccines, especially PCSK9Qβ-003 vaccine, developed high titer IgG antibodies against PCSK9. PCSK9Qβ-003 vaccine obviously decreased plasma total cholesterol in both Balb/c mice and LDLR
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, powder