- Characterization of a monoclonal antibody specific for human peripheral myelin protein 22 and its use in immunohistochemical studies of the fetal and adult nervous system.
Characterization of a monoclonal antibody specific for human peripheral myelin protein 22 and its use in immunohistochemical studies of the fetal and adult nervous system.
Journal of the peripheral nervous system : JPNS (2007-03-22)
Norman A Gregson, Gaiping Zhang, Jane Pritchard, Aiping Wang, Lara Sanvito, Adrian C Hayday, Richard A C Hughes
PMID17374097
ABSTRACT
We produced a mouse monoclonal antibody using cDNA and peptide immunization against the putative second extra-cellular domain of human peripheral myelin protein 22 (PMP22). It reacted specifically with human PMP22 and not with other human myelin proteins and did not react with bovine, rat, or mouse PMP22. The antibody stained the compact myelin of human peripheral nerve motor and sensory axons and did not stain central nervous system tissue. PMP22 reactivity was detected in the spinal roots of the human fetus at 19-20 weeks of gestation. The staining pattern of the PMP22 antibody resembled that of a monoclonal antibody directed against the myelin protein zero.