Merck
  • NMDA receptor mediates proliferation and CREB phosphorylation in postnatal Müller glia-derived retinal progenitors.
NMDA receptor mediates proliferation and CREB phosphorylation in postnatal Müller glia-derived retinal progenitors.
Molecular vision (2009-04-15)
Mónica Ramírez, Mónica Lamas
ABSTRACT
Postnatal retinal Müller glia are considered to be retinal progenitors as they retain the ability to dedifferentiate, proliferate, and differentiate to new retinal glia and neurons after injury. The proliferation and differentiation processes are coordinated by several extrinsic factors and neurotransmitters, including glutamate. Thus, the appropriate numbers and proportions of the different cell types are generated to form a functional retina during development and during injury repair. Here we analyze the changes in the proliferation of postnatal Müller glia-derived progenitors after activation of the N-methyl-D-aspartate (NMDA) glutamate receptors. Müller glia-derived progenitor cell cultures were characterized by immunocytochemistry with antibodies against the NR1 subunit of the NMDA receptor and the progenitor cell marker nestin. The effect of glutamate receptor agonists and antagonists on cell proliferation was analyzed by BrdU incorporation or Ki67 immunostaining, cell counting, and by immunolabeling of phosphorylated cAMP response element binding protein (P-CREB) transcription factor. The effect of NMDA receptor activation was analyzed in vivo by P-CREB immunohistochemistry in retinal sections of Long-Evans NMDA injected rats. We show that NMDA receptor activation significantly increases the proliferation rate of Müller-glia derived progenitor cells and that this increase can be blocked by NMDA receptor antagonists. Furthermore, we show that CREB phosphorylation is induced in NMDA-treated Müller-glia derived progenitor cells in culture and that specific pharmacological inhibition of CREB phosphorylation results in a decreased number of proliferating cells. We confirmed the relevance of these observations by the analysis of retinal sections after NMDA injection in vivo where immunoreactivity to phosphorylated CREB is also increased after treatment. In the present study we show that NMDA receptor activation induces postnatal Müller glia-derived retinal cell progenitor proliferation and transcription factor CREB phosphorylation both in culture and in vivo. The identification of the molecular determinants of mature retinal progenitors such as transcription factor CREB and NMDA receptor-induced players should facilitate the control of growth and manipulation of progenitor cell cultures and the possible identification of the molecular mechanisms involved in progenitor self-renewal.
Product Description
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Roche
5-Bromo-2′-deoxy-uridine Labeling and Detection Kit I, sufficient for ≤100 tests, kit of 1 (5 components), suitable for immunofluorescence