  • Search Results
  • Antinociceptive, anti-inflammatory and smooth muscle relaxant activities of the pyrrolo[3,4-d]pyridazinone derivatives: Possible mechanisms of action.
Antinociceptive, anti-inflammatory and smooth muscle relaxant activities of the pyrrolo[3,4-d]pyridazinone derivatives: Possible mechanisms of action.
Pharmacology, biochemistry, and behavior (2015-04-08)
Szczepan Mogilski, Monika Kubacka, Aleksandra Redzicka, Grzegorz Kazek, Magdalena Dudek, Wiesław Malinka, Barbara Filipek
ABSTRACT
The aim of this study was to evaluate the analgesic as well as anti-inflammatory activities of the new pyrrolo[3,4-d]pyridazinone derivatives. Moreover, the present study attempted to assess some of the mechanisms involved in the pharmacological activity of these compounds. In the previous studies it was shown that these compounds were highly active in the phenylbenzoquinone-induced 'writhing syndrome' test and had much lower activity in the hot plate, which indicates that mainly peripheral mechanisms of analgesia are involved in their effects. In these extended studies the analgesic activity of two tested compounds (4c, 4f) was confirmed in some animal models of pain. The studied compounds showed a significant and dose-related antinociceptive effect in the models of pain induced by formalin, capsaicin and glutamic acid. Both compounds decreased the edema formation and one of them (4c) attenuated mechanical hyperalgesia in carrageenan-induced paw inflammation in rats. Furthermore, both compounds inhibited cell migration, plasma exudation and nociceptive reaction in zymosan A-induced mouse peritonitis. In the subsequent studies, including experiments on isolated organs (ileum, trachea, aorta), radioligand assays and biochemical tests, it was demonstrated that analgesic and anti-inflammatory effects of the investigated structures are largely due to their competitive antagonism for histamine H1 receptor. The influence on the level of cAMP in inflammatory cells (shown in RAW 264.7 macrophages) and subsequent inhibition of cytokine (TNFα, IL-1β) release can also be one of the important mechanisms of their action. Moreover some additional mechanisms may also be involved in the eventual analgesic effect of tested pyrrolo[3,4-d]pyridazinone derivatives.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Antazoline hydrochloride
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG