  • Search Results
  • Revealing praziquantel molecular targets using mass spectrometry imaging: an expeditious approach applied to Schistosoma mansoni.
Revealing praziquantel molecular targets using mass spectrometry imaging: an expeditious approach applied to Schistosoma mansoni.
International journal for parasitology (2015-03-31)
Mônica Siqueira Ferreira, Rosimeire Nunes de Oliveira, Diogo Noin de Oliveira, Cibele Zanardi Esteves, Silmara Marques Allegretti, Rodrigo Ramos Catharino
ABSTRACT
Finding specific molecular targets and the mechanism of action of praziquantel in the treatment of schistosomiasis remains a challenging task. Our efforts were focused on obtaining further information on worm composition before and after exposure to praziquantel in the treatment of schistosomiasis to elucidate the potential sites of action of this drug. Evidence indicates that the lipid bilayer is changed by treatment with praziquantel. Following this rationale, we employed a mass spectrometry imaging-based approach that helped to characterise lipids in specific locations, which are directly involved in the biochemical pathways of the BH strain of Schistosoma mansoni, as well as differentiating the molecular response that each worm sex presents in vivo. Our findings demonstrated significant differences between the chemical markers found in adult worms before and after praziquantel exposure, especially in phospholipids, which were predominantly identified as chemical markers in all samples. Results also indicate that distinct molecular pathways in both male and female worms could be differentially affected by praziquantel treatment. These data shine new light on the mechanism of action of praziquantel, taking a further step towards its full understanding.
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles