Merck
Fibrosis is a common outcome following total knee arthroplasty.
Scientific reports (2015-11-11)
Nicole Abdul, David Dixon, Andrew Walker, Joanna Horabin, Nick Smith, David J Weir, Nigel T Brewster, David J Deehan, Derek A Mann, Lee A Borthwick
ABSTRACT
Total knee arthroplasty (TKA) is one of the most successful orthopaedic procedures that alleviates pain and restores function in patients with degenerative knee joint diseases. Arthrofibrosis, abnormal scarring in which dense fibrous tissue prevents normal range of motion, develops in ~3-10% of TKA patients. No prophylactic intervention is available and treatment is restricted to aggressive physiotherapy or revision surgery. Tissue was collected from patients undergoing primary (n = 30) or revision (n = 27) TKA. Revision patients were stratified as non-arthrofibrotic and arthrofibrotic. Tissue was macroscopically and histologically compared to improve our understanding of the pathophysiology of arthrofibrosis. Macroscopically, tissue from primary TKA presents as homogenous, fatty tissue whereas tissue from revision TKA presents as dense, pigmented tissue. Histologically, there was dramatic tissue remodelling, increased collagen deposition and increased (myo)fibroblast staining in tissue from revision TKA. Significantly, tissue architecture was similar between revision patients regardless of clinically diagnosis. There are significant differences in architecture and composition of tissue from revision TKA over primary TKA. Surprisingly, whether revision TKA were clinically diagnosed as arthrofibrotic or non-arthrofibrotic there were still significant differences in fibrotic markers compared to primary TKA suggesting an ongoing fibrotic process in all revision knees.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, SAJ first grade, 9.5-10.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 3% in ethanol
Sigma-Aldrich
Hydrochloric acid, suitable for arsenic determination, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ super special grade, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.1 M
Sigma-Aldrich
Hydrochloric acid, suitable for determination of toxic metals, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M