  • Search Results
  • Synthesis, cytotoxicity and antiviral evaluation of new series of imidazo[4,5-g]quinoline and pyrido[2,3-g]quinoxalinone derivatives.
Synthesis, cytotoxicity and antiviral evaluation of new series of imidazo[4,5-g]quinoline and pyrido[2,3-g]quinoxalinone derivatives.
European journal of medicinal chemistry (2015-10-20)
Irene Briguglio, Roberta Loddo, Erik Laurini, Maurizio Fermeglia, Sandra Piras, Paola Corona, Paolo Giunchedi, Elisabetta Gavini, Giuseppina Sanna, Gabriele Giliberti, Cristina Ibba, Pamela Farci, Paolo La Colla, Sabrina Pricl, Antonio Carta
ABSTRACT
Linear aromatic N-tricyclic compounds with promising antiviral activity and minimal cytotoxicity were prepared and analyzed in the last years. Specifically, the pyrido[2,3-g]quinoxalinone nucleus was found endowed with high potency against several pathogenic RNA viruses as etiological agents of important veterinary and human pathologies. Following our research program on new antiviral agents we have designed, synthesized and assayed new series of imidazo[4,5-g]quinoline and pyrido[2,3-g]quinoxalinone derivatives. Lead compounds 1-4 were further modified to enhance their antiviral activity and reduce their cytotoxicity. Thus, different substituents were introduced on N atom at position 1 or the O atom at position 2 of the leads; contextually, several groups were inserted on the nitrogen atom at position 7 of diaminoquinoline intermediates. Title compounds were tested in cell-based assays for cytotoxicity and antiviral activity against RNA virus families containing single-stranded (either positive-sense (ssRNA+) or negative-sense (ssRNA-)), and double-stranded genomes (dsRNA), and against two representatives of DNA virus families. Some derivatives emerged as potential leads for further development as antiviral agents against some viruses of public health significance, such as RSV, Reo, BVDV and HCV. Particularly, compounds 4, 11b, 11c, 13c, 15a, 18 and 21 resulted active against BVDV at concentrations ranging from 1.3 to 5 μM. Compound 21 was also evaluated for its activity on the BVDV RdRp. Compound 4 was also tested as potential anti-HCV compound in a subgenomic replication assay. Molecular simulation results provided a molecular rationale for the anti-BVDV activity of these compounds.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Glutamine, JIS special grade, ≥99.0%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)