  • Search Results
  • Shikonin inhibits adipogenic differentiation via regulation of mir-34a-FKBP1B.
Shikonin inhibits adipogenic differentiation via regulation of mir-34a-FKBP1B.
Biochemical and biophysical research communications (2015-10-17)
Young Jin Jang, Chang Hwa Jung, Jiyun Ahn, So Young Gwon, Tae Youl Ha
ABSTRACT
Shikonin is a naturally occurring naphthoquinone pigment and a major constituent present in Lithospermum erythrorhizon. Since microRNAs (miRNAs) are one of the key post-transcriptional regulators of adipogenesis, their manipulation represents a potential new strategy to inhibit adipogenesis. Our aim was to investigate shikonin-dependent inhibition of adipogenesis with an emphasis on miRNA-related processes. Mir-34a increased during induced adipogenesis, and this was suppressed in the presence of shikonin. mRNA expression of FKBP1B, a suggested target of mir-34a according to bioinformatics studies, decreased during adipogenesis, but was recovered by shikonin treatment, which reversely correlated with mir-34a expression. A mir-34a inhibitor suppressed MDI-induced adipogenesis by blocking PPARγ and C/EBPα expression, while suppression of mir-34a recovered MDI-induced down-regulation of FKBP1B expression. A mir-34a mimic decreased FKBP1B mRNA expression in 3T3-L1 preadipocytes. We also observed that mir-34a bound directly to the 3'-untranslated region of FKBP1B. Finally, FKBP1B overexpression attenuated MDI-induced adipogenesis, PPARγ, and C/EBPα expression. These results suggest that mir-34a regulates adipogenesis by targeting FKBP1B expression. Our findings reveal that shikonin prevents adipogenesis by blocking the mir-34a-FKBP1B pathway which represents a promising potential target for preventing obesity.
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
1,2-Propanediol, LR, ≥99%
Sigma-Aldrich
Propylene glycol, FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications