  • Search Results
  • Reversal of P-glycoprotein overexpression by Ginkgo biloba extract in the brains of pentylenetetrazole-kindled and phenytoin-treated mice.
Reversal of P-glycoprotein overexpression by Ginkgo biloba extract in the brains of pentylenetetrazole-kindled and phenytoin-treated mice.
The Kaohsiung journal of medical sciences (2015-08-01)
Ce Zhang, Qing Fan, Shu-Liang Chen, Hui Ma
ABSTRACT
The purpose of this study was to investigate the combined effects of Ginkgo biloba extract and phenytoin (PHT) sodium as a dose regimen simulating the clinical treatment of patients with epilepsy, on P-glycoprotein (P-GP) overexpression in a pentylenetetrazole-kindled mouse model of epilepsy. Epilepsy was induced by intraperitoneal administration of pentylenetetrazole (40 mg/kg) for 7 days followed by intragastric administration of PHT (40 mg/kg) for 14 days. Thirty mice that developed seizures were randomly divided into three groups and administered PHT as well as the following treatments: saline (negative control); verapamil (20 mg/kg, positive control); and G. biloba (30 mg/kg). Seizure severity was recorded 30 minutes after treatment on Day 4 of drug administration, after which the mice were euthanized, and their brains isolated. Western blots and immunohistochemistry were performed to analyze the expression of P-GP and caspase-3, respectively, in the brain tissue. High-performance liquid chromatography was used to measure the concentrations of PHT in the brains of the treated mice. After 4 consecutive days of treatment, the seizure severity in the mice in the G. biloba extract group was more significantly reduced than the seizure severity in the saline control group, and a significant difference was observed between the G. biloba extract and verapamil control groups (p < 0.05). P-GP expression in the brain more significantly decreased in the mice treated with G. biloba extract and verapamil than it did in the saline-treated control group (p < 0.05). Compared with the saline-treated control group, the mice treated with G. biloba extract and verapamil showed significantly increased brain PHT concentrations (p < 0.05). Furthermore, caspase-3 expression in the brain tissue of the G. biloba extract group was significantly lower than that in the vehicle control group (p < 0.05); this finding demonstrated the neuroprotective effects of G. biloba. Therefore, this study showed that treatment with G. biloba extract in combination with PHT prevented the upregulation of P-GP expression in mice. Moreover, G. biloba extract decreased seizure severity in pentylenetetrazole-kindled/PHT-treated mice through a mechanism that might be related to the reduction of P-GP expression in the brain.
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Pyrrolidone hydrotribromide, 97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%