  • Search Results
  • The effect of various electrolyte cations on electrochemical performance of polypyrrole/RGO based supercapacitors.
The effect of various electrolyte cations on electrochemical performance of polypyrrole/RGO based supercapacitors.
Physical chemistry chemical physics : PCCP (2015-10-08)
Jianbo Zhu, Youlong Xu, Jie Wang, Jun Lin, Xiaofei Sun, Shengchun Mao
ABSTRACT
In this work, polypyrrole/graphene doped by p-toluenesulfonic is prepared as an active material for supercapacitors, and its capacitance performance is investigated in various aqueous electrolytes including HCl, LiCl, NaCl, and KCl with a concentration of 3 M, respectively. A rising trend of capacitance is observed according to the cationic mobility (Li(+) < Na(+) < K(+) < H(+)), which is due to its effect on the ionic conductivity, efficient ion/charge diffusion/exchange and relaxation time. On the other hand, long-term cycling stability is in the following order: KCl < NaCl < LiCl < HCl, corresponding to the decreasing tendency of cation size (K(+) > Na(+) > Li(+) > H(+)). The reason can be attributed to the fact that the insertion/de-insertion of large size cation brings a significant doping level decrease and an over-oxidation increase during the charging-discharging cycles. Hence, we not only obtain good capacitance performance (280.3 F g(-1) at 5 mV s(-1)), superior rate capability (225.8 F g(-1) at 500 mV s(-1)) and high cycling stability (92.0% capacitance retention after 10,000 cycles at 1 A g(-1)) by employing 3 M HCl as an electrolyte, but also reveal that the electrolyte cations have a significant effect on the supercapacitors' electrochemical performance.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O