  • Search Results
  • INPP4B Is a PtdIns(3,4,5)P3 Phosphatase That Can Act as a Tumor Suppressor.
INPP4B Is a PtdIns(3,4,5)P3 Phosphatase That Can Act as a Tumor Suppressor.
Cancer discovery (2015-04-18)
Satoshi Kofuji, Hirotaka Kimura, Hiroki Nakanishi, Hiroshi Nanjo, Shunsuke Takasuga, Hui Liu, Satoshi Eguchi, Ryotaro Nakamura, Reietsu Itoh, Noriko Ueno, Ken Asanuma, Mingguo Huang, Atsushi Koizumi, Tomonori Habuchi, Masakazu Yamazaki, Akira Suzuki, Junko Sasaki, Takehiko Sasaki
ABSTRACT
Inositol polyphosphate 4-phosphatase B (INPP4B) has been identified as a tumor suppressor mutated in human breast, ovary, and prostate cancers. The molecular mechanism underlying INPP4B's tumor-suppressive role is currently unknown. Here, we demonstrate that INPP4B restrains tumor development by dephosphorylating the PtdIns(3,4,5)P3 that accumulates in situations of PTEN deficiency. In vitro, INPP4B directly dephosphorylates PtdIns(3,4,5)P3. In vivo, neither inactivation of Inpp4b (Inpp4b(Δ/Δ)) nor heterozygous deletion of Pten (Pten(+/-)) in mice causes thyroid abnormalities, but a combination of these mutations induces malignant thyroid cancers with lung metastases. At the molecular level, simultaneous deletion of Inpp4b and Pten synergistically increases PtdIns(3,4,5)P3 levels and activates AKT downstream signaling proteins in thyroid cells. We propose that the PtdIns(3,4,5)P3 phosphatase activity of INPP4B can function as a "back-up" mechanism when PTEN is deficient, making INPP4B a potential novel therapeutic target for PTEN-deficient or PIK3CA-activated cancers. Although INPP4B expression is reduced in several types of human cancers, our work on Inpp4B-deficient mice provides the first evidence that INPP4B is a bona fide tumor suppressor whose function is particularly important in situations of PTEN deficiency. Our biochemical data demonstrate that INPP4B directly dephosphorylates PtdIns(3,4,5)P3.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%