Merck
Ginger extract ameliorates phosphamidon induced hepatotoxicity.
Indian journal of experimental biology (2015-11-10)
Suprabhat Mukherjee, Niladri Mukherjee, Prasanta Saini, Priya Roy, Santi P Sinha Babu
ABSTRACT
Organophosphorus (OP) compounds commonly used as pesticides in agriculture cause serious health problems to living beings. The present study enumerates the ameliorating effect of ginger extract (GE) against phosphamidon (PHO, an organophosphorus insecticide) induced hepatotoxicity. GE was prepared from dried ginger and characterized for compound profile and antioxidant activity. Eight groups of albino rats (n = 6) were treated with 1/5th lethal dose of PHO for 5-20 days. Out of the treated 8 groups, 4 were simultaneously fed with GE (1 mg/kg body wt.) along with PHO. Alterations in the levels of hepatocellular oxidative stress (OS) markers in the treated groups indicated an enhanced generation of reactive oxygen species (ROS) and oxidative stress (OS). Upregulation of apoptotic markers, DNA fragmentation and appearance of apoptotic nuclei suggested induction of apoptosis in the liver cell that was found to be attenuated after GE treatment. Moreover, no toxicity and mortality was observed up to 100 mg/kg dose of GE for 30 days in the rat model studied. Thus, GE can be considered as an effective, economical and safe extract to circumvent PHO-induced hepatotoxicity.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, SAJ first grade, 88.0-89.5%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%