  • Search Results
  • Preparation Method of Crack-free PVDF Microfiltration Membrane with Enhanced Antifouling Characteristics.
Preparation Method of Crack-free PVDF Microfiltration Membrane with Enhanced Antifouling Characteristics.
ACS applied materials & interfaces (2015-07-15)
Sahng Hyuck Woo, Ju Sung Lee, Hyun Ho Lee, Jinwon Park, Byoung Ryul Min
ABSTRACT
This study proposes a method to prepare a crack-free poly(vinylidene fluoride) (PVDF) microfiltration (MF) membrane with enhanced antifouling property. In the study, blending 4% poly(vinylidene fluoride)-graft-poly(sulfopropyl methacrylate) (PVDF-g-PSPMA) and 1.5% potassium perchlorate (KClO4) led to crack prevention during membrane preparation via nonsolvent induced phase separation (NIPS) when compared with blending with 4% PVDF-g-PSPMA only (without KClO4). The resulting crack-free membrane (A3) had both smooth surface structure and hydrophilicity in comparison with pristine PVDF membrane (A1). In addition, blending with PVDF-g-PSPMA and KClO4 also allowed the A3 membrane to exhibit uniform pore size distribution (PSD) and smooth surface structure, compared with PVDF membrane commercially available from company "M" in Germany. The aforementioned properties led to antifouling characteristics in the crack-free membrane (A3). According to flux performances, flux recovery and cumulative permeate volume (between 120 and 240 min) of crack-free membrane (A3) were 11.41 and 17.41% superior to those of commercial membrane, respectively.
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 0.02 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide, ≥96.0%, for determination of nitrogen
Sigma-Aldrich
Sodium hydroxide solution, 0.5 M
Sigma-Aldrich
Sodium hydroxide solution, 2 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 20 % (w/v)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O