  • Search Results
  • Single-step multiresidue determination of ten multiclass veterinary drugs in pork, milk, and eggs using liquid chromatography with tandem mass spectrometry.
Single-step multiresidue determination of ten multiclass veterinary drugs in pork, milk, and eggs using liquid chromatography with tandem mass spectrometry.
Journal of separation science (2015-06-03)
Jin-A Park, Dan Zhang, Dong-Soon Kim, Seong-Kwan Kim, Kyeong-Su Cho, Dana Jeong, Jae-Han Shim, Jin-Suk Kim, A M Abd El-Aty, Ho-Chul Shin
ABSTRACT
A multiclass, multiresidue determination method is reported for the detection of ten veterinary drugs, including scopolamine, metoclopramide, acriflavine, berberine, tripelennamine, diphenhydramine, acrinol, triamcinolone, loperamide, and roxithromycin in pork, milk, and eggs. The method involves a simple extraction using 0.1% formic acid in acetonitrile, followed by defatting with n-hexane, centrifugation, and filtration prior to liquid chromatography with tandem mass spectrometric analysis. As ion suppression and enhancement effects are reported, matrix-matched calibrations are used for quantification, with determination coefficients ≥0.9765. For the majority of the tested analytes, the intra- and interday accuracy (expressed as recovery %) range from 70.6 to 94.6% and from 70.1 to 93.3%, respectively, and the precision (expressed as relative standard deviation) ranges from 0.5 to 19.8% and from 2.8 to 18.4% in all matrices. The limits of quantification range between 0.5 and 10 ng/g. The validated tandem mass spectrometry method is successfully applied to market samples; the target analytes are not detected in any of the tested samples. In terms of accuracy, no extract cleanup is deemed necessary. The developed method is feasible for the simultaneous detection of the tested analytes in pork, milk, and eggs.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Roxithromycin, 95.0-102.0% (HPLC)
Sigma-Aldrich
Triamcinolone
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acriflavine
Sigma-Aldrich
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acriflavine, suitable for fluorescence, BioReagent, ≥90% (AT)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Formic acid, SAJ first grade, 88.0-89.5%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC