  • Search Results
  • Investigating the Potential Effect of Commiphora myrrha on the Pharmacokinetics of Theophylline, a Narrow Therapeutic Index Drug.
Investigating the Potential Effect of Commiphora myrrha on the Pharmacokinetics of Theophylline, a Narrow Therapeutic Index Drug.
Drug research (2014-07-06)
F I Al-Jenoobi, A Ahad, M Raish, A M Al-Mohizea, M A Alam
ABSTRACT
The present study was conducted to investigate the effect of commonly used herb Commiphora myrrha on the pharmacokinetic profile of theophylline (narrow therapeutic index drug) in rabbits. In the experimental groups, theophylline (16 mg/kg) was given orally to the rabbits. Where aqueous saline suspension of Commiphora myrrha (176 mg/kg, p.o.), was given to the rabbits and the blood samples were withdrawn at different time intervals (0, 0.5, 1, 1.5, 2, 3, 4, 6, 8, 12, 24 and 36 h) from marginal ear vein after dosing and theophylline in plasma was analyzed by HPLC method. It was observed that there a significant differences in the Cmax, AUC, AUMC, t1/2, and MRT of theophylline when coadministered with Commiphora myrrha which indicate that the herb affect the metabolism and elimination when coadministered with theophylline. Our results suggested that concurrent use of investigated herb alters the pharmacokinetics of theophylline. Confirmation of these results in human studies will warrant changes in theophylline dose or frequency when coadministered with herb under consideration.
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid