  • Search Results
  • Exposure to sub-chronic unpredictable stress accounts for antidepressant-like effects in hamsters treated with BDNF and CNQX.
Exposure to sub-chronic unpredictable stress accounts for antidepressant-like effects in hamsters treated with BDNF and CNQX.
Brain research bulletin (2015-09-27)
Raffaella Alò, Maria Mele, Gilda Fazzari, Ennio Avolio, Marcello Canonaco
ABSTRACT
Recent evidences indicate that cerebral neurotrophic factors like vascular endothelial growth factor plus signaling pathways of the glutamatergic neuroreceptor system (L-Glu) are determinant modulators of depression-like states. In the present study, the type of interaction(s) exerted by the AMPAergic antagonist, 6-cyano-7-nitroquinoxalin-2,3-dione (CNQX) and the brain derived neurotrophic factor (BDNF) on depression-like behaviors in hamsters (Mesocricetus auratus) were investigated. Sub-chronic administration of BDNF in the hippocampal dentate gyrus (DG) of stressed hamsters was responsible for very evident (p<0.001) sucrose consumption along with notably elevated swimming bouts and reduced immobility states in the forced swim test (FST). Meanwhile, CNQX displayed evident anxiolytic actions in the elevated plus maze (EPM) as shown by marked (p<0.01) increases of movements to and from both arms. Interestingly cerebral neurodegeneration events, which are viewed during depression states, were reduced following treatment with both compounds. Contextually, marked mRNA expression levels of the BDNF receptor (tropomyosin-related kinase B; TrkB) were detected in DG and the oriens-pyramidalis of HIP (Or-Py) while a moderate (p<0.05) up-regulation was registered in the amygdalar central nucleus (CeA) and the hypothalamic ventromedial nucleus (VMH) of hamsters treated with BDNF. Similarly, this treatment caused moderate increases of the major stress protein (Hsp70) in DG and Or-Py. Conversely, while CNQX induced similar TrkB expression levels, it instead accounted for a moderate reduction of Hsp70 mRNAs in the same brain areas. Overall these results support crucial roles played by BDNF and AMPAergic neurosignaling mechanisms during distinct adaptive responses of depression- and anxiety-like states in hamsters.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis