  • Search Results
  • Vitamin Bc -Bearing Hydrophilic Photosensitizer Conjugate for Photodynamic Cancer Theranostics.
Vitamin Bc -Bearing Hydrophilic Photosensitizer Conjugate for Photodynamic Cancer Theranostics.
Macromolecular bioscience (2015-04-02)
Jiyoung Kim, Kyoung Sub Kim, Sin-jung Park, Kun Na
ABSTRACT
The accurate diagnosis and proper therapy for cancer are essential to improve the success rate of cancer treatment. Here, we demonstrated that the vitamin Bc -bearing hydrophilic photosensitizer conjugate folic acid-polyethylene glycol-pheophorbideA (FA-PEG-PheoA) has been synthesized for the intracellular diagnosis and photodynamic therapy of a tumor. The synthesized vitamin Bc -bearing hydrophilic photosensitizer conjugate has been characterized for the folic acid receptor expressing the ability to target tumor cells, which is facilitated by the chemical conjugation with folic acid. The vitamin Bc -bearing hydrophilic photosensitizer conjugate internalization mechanism was identified through a competitive inhibition test with free folic acid. We optimized the laser-sensitive, cytotoxicity changeable, vitamin Bc -bearing hydrophilic photosensitizer conjugate concentration, which is non-cytotoxic under normal conditions and specifically cytotoxic toward cancer cells (maximum 69.15%) under laser irradiation conditions used for theranostic agents. The cancer therapeutic and diagnosis effects of synthesized conjugate were confirmed in MDA-MB-231 cells and MDA-MB-231-bearing mice. As a result, the vitamin Bc -bearing hydrophilic photosensitizer conjugate exhibited a highly photodynamic therapeutic effect, which enabled the selective detection of a folic acid receptor expressing cancer using optical imaging.
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium bromide, JIS special grade, 99.0-100.2%
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Potassium bromide, SAJ first grade, 99.0-101.0%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Potassium bromide, AR, ≥99.5%
Sigma-Aldrich
Potassium bromide, LR, ≥99%
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, AR, ≥99.5%
Sigma-Aldrich
Methanol, LR, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
DCC, 99%