  • Search Results
  • Determination of phytochemicals, antioxidant activity and total phenolic content in Andrographis paniculata using chromatographic methods.
Determination of phytochemicals, antioxidant activity and total phenolic content in Andrographis paniculata using chromatographic methods.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-06-09)
Marzanna Kurzawa, Anna Filipiak-Szok, Ewa Kłodzińska, Edward Szłyk
ABSTRACT
Antioxidant activity, total phenolics content and selected phytochemicals (alkaloids and andrographolides) were determined in Andrographis paniculata and in dietary supplements containing this plant. Antioxidant activity was measured by FRAP, CUPRAC and DPPH procedures and ranged from 503.36 to 6164.09μmol TE/100g d.m. depending on methods, part of plant and kind of dietary supplement. The total phenolics (175.13-1723.79mg GAE/100g) and andrographolides content (19.44-85.13mg/g) in the studied samples were correlated with antioxidant activities determined by CUPRAC, FRAP and DPPH (r>0.95, p<0.05 level). Purine alkaloids: caffeine, theobromine, theophylline and indole alkaloids: harmine, harmane, harmol, yohimbine, brucine and strychnine were detected in the studied samples by different chromatographic techniques (HPLC-DAD, LC-MS/MS, GC-MS). The total alkaloids content in APs-roots and APs-leaves varies from 50.71±0.36mg/g d.m. to 78.71±0.48mg/g d.m., respectively, whereas for dietary supplements (Pn and DK) TAC was found between 19.52±0.15mg/g and 22.18±0.15mg/g d.m.. The highest concentration of andrographolides was found in A. paniculata leaves, whereas the lowest in dietary supplement Pn. Moreover principal component analysis, cluster analysis and one-way ANOVA follow by Duncan's tests were also performed.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Strychnine
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
SAFC
Acetic acid, glacial
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis