Merck
Functional properties of submucosal venules in the rat stomach.
Pflugers Archiv : European journal of physiology (2014-07-30)
Retsu Mitsui, Hikaru Hashitani
ABSTRACT
Venules in the stomach may have intrinsic properties for maintaining active microcirculation drainage even during gastric filling. Properties of spontaneous and nerve-mediated activity of submucosal venules in the rat stomach were investigated. Changes in vasodiameter and intracellular Ca(2+) in venular smooth muscle cells (SMCs) were monitored by video tracking and Fluo-8 Ca(2+) imaging, respectively. Venular SMCs developed synchronous spontaneous Ca(2+) transients and corresponding rhythmic constrictions of the venules. Nominally Ca(2+)-free solution or an L-type Ca(2+) channel blocker (1 μM nifedipine) disrupted the Ca(2+) transient synchrony and abolished spontaneous constrictions. Spontaneous constrictions were also prevented by inhibitors of sarcoplasmic reticulum Ca(2+)-ATPase (10 μM cyclopiazonic acid (CPA)), IP3 receptors (100 μM 2-APB) or Ca(2+)-activated Cl(-) channels (100 μM niflumic acid). Transmural nerve stimulation (TNS) induced a long-lasting venular constriction that was abolished by α-adrenoceptor antagonist (1 μM phentolamine), while TNS evoked a sympathetic transient constriction of arterioles that was abolished by a combination of phentolamine and a P2 purinoceptor antagonist (10 μM pyridoxal-phosphate-6-azophenyl-2',4'-disulfonic acid (PPADS)). Consistently, P2X1 purinoceptor immunoreactivity was detected in arteriolar but not venular SMCs. Primary afferent nerve stimulation (300 nM capsaicin) caused a venular dilatation by releasing calcitonin gene-related peptide. Thus, Ca(2+) release from the sarcoplasmic reticulum may play a fundamental role in the generation of spontaneous Ca(2+) transients, while electrical coupling amongst venular SMCs via L-type Ca(2+) channel activation appears to be critical for Ca(2+) transient synchrony as well as spontaneous contractions. Sympathetic venular constrictions appear to be exclusively mediated by noradrenaline due to the lack of P2X1 receptor in venular SMCs.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate, ≥80% (elemental analysis), powder
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Niflumic acid
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Tetracaine, meets USP testing specifications
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)