  • Search Results
  • Effects of dietary onion (Allium cepa) powder on growth, innate immune response and hemato-biochemical parameters of beluga (Huso huso Linnaeus, 1754) juvenile.
Effects of dietary onion (Allium cepa) powder on growth, innate immune response and hemato-biochemical parameters of beluga (Huso huso Linnaeus, 1754) juvenile.
Fish & shellfish immunology (2015-06-13)
Raza Akrami, Ahmad Gharaei, Majid Razeghi Mansour, Ali Galeshi
ABSTRACT
The present study was aimed at determining the effects of dietary onion powder on growth, innate immune response and hemato-biochemical parameters of beluga juvenile (Huso huso). Basal diets containing onion powder 0 (control), 0.5 and 1% of feed were fed to beluga juvenile. At the end of the experiment, the highest weight gain (WG%) and specific growth rate (SGR) was observed in group fed with 1% onion (P < 0.05). There were no significant difference (P > 0.05) about feed conversion ratio (FCR) in treatment groups that fed diets containing various levels of onion powder. After 8 weeks, serum lysozyme activity, superoxide dismutase activity (SOD), respiratory burst activity and serum total immunoglobulin (Ig) showed a significant increase in treatment group with 1% onion powder compared to other groups (P < 0.05). The group fed 1% onion showed a significantly increases in the number of erythrocytes (RBC), leucocyte (WBC), haematocrit (Hct) levels compared to the control group (P < 0.05). Haemoglobin, monocyte, lymphocyte and neutrophil had no significant change (P > 0.05) in treatment groups and control. The analysis of AST and LDH levels showed a significant decrease in 1% onion compared to the control and 0.5% onion diet (P < 0.05), while ALT and ALP levels were not influenced (P > 0.05). The blood glucose, total protein, triglyceride, cholesterol, albumin and globulin levels were lower in treated groups compared with the control (P < 0.05). The results of this study demonstrated that dietary onion powder could be an improvement in growth, hematological parameters and immune function of beluga juvenile.
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)