  • Search Results
  • Improvements on neuromuscular performance with caffeine ingestion depend on the time-of-day.
Improvements on neuromuscular performance with caffeine ingestion depend on the time-of-day.
Journal of science and medicine in sport (2014-05-13)
Ricardo Mora-Rodríguez, Jesús G Pallarés, José María López-Gullón, Álvaro López-Samanes, Valentín E Fernández-Elías, Juan F Ortega
ABSTRACT
To determine whether the ergogenic effects of caffeine ingestion on neuromuscular performance are similar when ingestion takes place in the morning and in the afternoon. Double blind, cross-over, randomized, placebo controlled design. Thirteen resistance-trained males carried out bench press and full squat exercises against four incremental loads (25%, 50%, 75% and 90% 1RM), at maximal velocity. Trials took place 60 min after ingesting either 6 mg kg(-1) of caffeine or placebo. Two trials took place in the morning (AMPLAC and AMCAFF) and two in the afternoon (PMPLAC and PMCAFF), all separated by 36-48 h. Tympanic temperature, plasma caffeine concentration and side-effects were measured. Plasma caffeine increased similarly during AMCAFF and PMCAFF. Tympanic temperature was lower in the mornings without caffeine effects (36.7±0.4 vs. 37.0±0.5°C for AM vs. PM; p<0.05). AMCAFF increased propulsive velocity above AMPLAC to levels similar to those found in the PM trials for the 25%, 50%, 75% 1RM loads in the SQ exercise (5.4-8.1%; p<0.05). However, in the PM trials, caffeine ingestion did not improve propulsive velocity at any load during BP or SQ. The negative side effects of caffeine were more prevalent in the afternoon trials (13 vs. 26%). The ingestion of a moderate dose of caffeine counteracts the muscle contraction velocity declines observed in the morning against a wide range of loads. Caffeine effects are more evident in the lower body musculature. Evening caffeine ingestion not only has little effect on neuromuscular performance, but increases the rate of negative side-effects reported.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG