  • Search Results
  • Divergent synthesis of 4,6-diarylated pyridin-2(1H)-ones from chalcones: novel access to 2,4,6-triaryl pyridines.
Divergent synthesis of 4,6-diarylated pyridin-2(1H)-ones from chalcones: novel access to 2,4,6-triaryl pyridines.
Organic & biomolecular chemistry (2015-05-01)
Rajni Khajuria, Prakash Kannaboina, Kamal K Kapoor, Annah Gupta, Gaurav Raina, Amanpreet Kaur Jassal, Love Karan Rana, Maninder S Hundal, Parthasarathi Das
ABSTRACT
A wide range of 4,6-diarylated/heterylated pyridin-2(1H)-one derivatives were synthesized in good to excellent yields from 1,3-diarylated/heterylated-2-propen-1-ones (chalcones) in one pot under metal and base-free conditions. This domino reaction suggests a novel mechanism comprising of Michael addition followed by amination, subsequent intramolecular amidation and finally dehydronitrosation. The usefulness of the designed 4,6-diarylated/heterylated pyridin-2(1H)-one derivatives has further been demonstrated by synthesizing medicinally important 2,4,6-triaryl/heteryl pyridines via Pd-catalyzed cross-coupling reaction.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous