  • Search Results
  • Mechanisms of the sarcoplasmic reticulum Ca2+ release induced by P2X receptor activation in mesenteric artery myocytes.
Mechanisms of the sarcoplasmic reticulum Ca2+ release induced by P2X receptor activation in mesenteric artery myocytes.
Pharmacological reports : PR (2014-06-07)
Khrystyna Yu Sukhanova, Oleksandr M Thugorka, Vitali A Bouryi, Maksym I Harhun, Dmitri V Gordienko
ABSTRACT
ATP is one of the principal sympathetic neurotransmitters which contracts vascular smooth muscle cells (SMCs) via activation of ionotropic P2X receptors (P2XRs). We have recently demonstrated that contraction of the guinea pig small mesenteric arteries evoked by stimulation of P2XRs is sensitive to inhibitors of IP3 receptors (IP3Rs). Here we analyzed contribution of IP3Rs and ryanodine receptors (RyRs) to [Ca(2+)]i transients induced by P2XR agonist αβ-meATP (10 μM) in single SMCs from these vessels. The effects of inhibition of L-type Ca(2+) channels (VGCCs), RyRs and IP3Rs (5 μM nicardipine, 100 μM tetracaine and 30 μM 2-APB, respectively) on αβ-meATP-induced [Ca(2+)]i transients were analyzed using fast x-y confocal Ca(2+) imaging. The effect of IP3R inhibition on the [Ca(2+)]i transient was significantly stronger (67 ± 7%) than that of RyR inhibition (40 ± 5%) and was attenuated by block of VGCCs. The latter indicates that activation of VGCCs is linked to IP3R-mediated Ca(2+) release. Immunostaining of RyRs and IP3Rs revealed that RyRs are located mainly in deeper sarcoplasmic reticulum (SR) while sub-plasma membrane (PM) SR elements are enriched with type 1 IP3Rs. This structural peculiarity makes IP3Rs more accessible to Ca(2+) entering the cell via VGCCs. Thus, IP3Rs may serve as an "intermediate amplifier" between voltage-gated Ca(2+) entry and RyR-mediated Ca(2+) release. P2X receptor activation in mesenteric artery SMCs recruits IP3Rs-mediated Ca(2+) release from sub-PM SR, which is facilitated by activation of VGCCs. Sensitivity of IP3R-mediated release to VGCC antagonists in vascular SMCs makes this mechanism of special therapeutic significance.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
U-73343
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard