  • Search Results
  • FOXO1 is required for binding of PR on IRF4, novel transcriptional regulator of endometrial stromal decidualization.
FOXO1 is required for binding of PR on IRF4, novel transcriptional regulator of endometrial stromal decidualization.
Molecular endocrinology (Baltimore, Md.) (2015-01-15)
Yasmin M Vasquez, Erik C Mazur, Xilong Li, Ramakrishna Kommagani, Lichun Jiang, Rui Chen, Rainer B Lanz, Ertug Kovanci, William E Gibbons, Francesco J DeMayo
ABSTRACT
The forkhead box O1A (FOXO1) is an early-induced target of the protein kinase A pathway during the decidualization of human endometrial stromal cells (HESCs). In this study we identified the cistrome and transcriptome of FOXO1 and its role as a transcriptional regulator of the progesterone receptor (PR). Direct targets of FOXO1 were identified by integrating RNA sequencing with chromatin immunoprecipitation followed by deep sequencing. Gene ontology analysis demonstrated that FOXO1 regulates a subset of genes in decidualization such as those involved in cancer, p53 signaling, focal adhesions, and Wnt signaling. An overlap of the FOXO1 and PR chromatin immunoprecipitation followed by deep sequencing intervals revealed the co-occupancy of FOXO1 in more than 75% of PR binding intervals. Among these intervals were highly enriched motifs for the interferon regulatory factor member 4 (IRF4). IRF4 was determined to be a genomic target of both FOXO1 and PR and also to be differentially regulated in HESCs treated with small interfering RNA targeting FOXO1 or PR prior to decidualization stimulus. Ablation of FOXO1 was found to abolish binding of PR to the shared binding interval downstream of the IRF4 gene. Finally, small interfering RNA-mediated ablation of IRF4 was shown to compromise morphological transformation of decidualized HESCs and to attenuate the expression of the decidual markers IGFBP1, PRL, and WNT4. These results provide the first evidence that FOXO1 is functionally required for the binding of PR to genomic targets. Most notably, FOXO1 and PR are required for the regulation of IRF4, a novel transcriptional regulator of decidualization in HESCs.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SAFC
BIS-TRIS
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Medroxyprogesterone acetate, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, LR, ≥99%
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, AR, ≥99.5%
Sigma-Aldrich
Sodium chloride, LR, ≥99.5%
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet