  • Search Results
  • Controlled 3D culture in Matrigel microbeads to analyze clonal acinar development.
Controlled 3D culture in Matrigel microbeads to analyze clonal acinar development.
Biomaterials (2015-03-31)
Monika E Dolega, Fabien Abeille, Nathalie Picollet-D'hahan, Xavier Gidrol
ABSTRACT
3D culture systems are a valuable tool for modeling morphogenesis and carcinogenesis of epithelial tissue in a structurally appropriate context. We present a novel approach for 3D cell culture based on a flow-focusing microfluidic system that encapsulates epithelial cells in Matrigel beads. As a model we use prostatic and breast cells and assay for development of acini, polarized cellular spheres enclosing lumen. Each individual bead on average acts as a single 3D cell culture compartment generating one acinus per bead. Compared to standard protocols microfluidics provides increased control over the environment leading to more a uniform acini population. The increased facility of bead manipulation allowed us to isolate single cells which are self-sufficient to fully develop into acini in presence of Matrigel. Furthermore, combination of our microfluidic approach with large particle FACS opens new avenues in high throughput screening on single acini or spheroids.
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard