  • Search Results
  • Soy phosphatidylinositol containing nanoparticle prolongs hemostatic activity of B-domain deleted factor VIII in hemophilia A mice.
Soy phosphatidylinositol containing nanoparticle prolongs hemostatic activity of B-domain deleted factor VIII in hemophilia A mice.
Journal of pharmaceutical sciences (2014-04-05)
Krithika A Shetty, Matthew P Kosloski, Donald E Mager, Sathy V Balu-Iyer
ABSTRACT
Factor VIII (FVIII) replacement therapy in hemophilia A (HA) is complicated by a short half-life and high incidence of inhibitory antibody response against the protein. Phosphatidylinositol (PI) containing lipidic nanoparticles have previously been shown to reduce the immunogenicity and prolong the half-life of full length FVIII. It has not been established whether this prolongation in half-life improves hemostatic efficacy and whether this approach could be extended to the B-domain deleted form of FVIII (BDD FVIII). In the current study, we evaluated the pharmacokinetics (PK), hemostatic efficacy, and immunogenicity of BDD FVIII associated with PI nanoparticles (PI-BDD FVIII) in HA mice. Comparative human PK was predicted using an "informed scaling" approach. PI-BDD FVIII showed an approximate 1.5-fold increase in terminal half-life compared with free BDD FVIII following i.v. bolus doses of 40 IU/kg. PI-BDD FVIII-treated animals retained hemostatic efficacy longer than the free FVIII-treated group in a tail vein transection model of hemostasis. PI association reduced the development of inhibitory and binding antibodies against BDD FVIII after a series of i.v. injections. The combined improvements in circulating half-life and hemostatic efficacy could significantly prolong the time above clinically established therapeutic thresholds of prophylactic FVIII replacement therapy in humans.
Product Description
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
D-(+)-Glucose, AR, anhydrous
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, analytical standard
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%