  • Search Results
  • Clinicopathological significance of 'subtypes with stem-cell feature' in combined hepatocellular-cholangiocarcinoma.
Clinicopathological significance of 'subtypes with stem-cell feature' in combined hepatocellular-cholangiocarcinoma.
Liver international : official journal of the International Association for the Study of the Liver (2014-04-10)
Motoko Sasaki, Hirohide Sato, Yuko Kakuda, Yasunori Sato, Joon Hyuk Choi, Yasuni Nakanuma
ABSTRACT
Combined hepatocellular-cholangiocarcinoma (cHC-CC), a malignant liver tumour with poor prognosis, is composed of hepatocellular carcinoma (HCC), cholangiocarcinoma (CC) and diverse components with intermediate features between HCC and CC, which correspond to hepatic progenitor cells. According to the WHO classification 2010, we surveyed the prevalence and clinicopathological significance of each subtype with stem-cell features [SC subtype; typical subtype (TS), intermediate cell subtype (INT) and cholangiolocellular type (CLC)] in cHC-CC and HCC. Sixty-two patients with cHC-CC (19 women and 43 men) and 26 patients with HCC (all men) were examined. The prevalence of each component was histologically assessed with assistance of mucin and immunohistochemical stainings. SC subtypes were observed in all cHC-CCs in various amount and combination. The prevalence of each SC subtype in cHC-CC was as follows: TS, 10 (16.1%); INT, 53 (83.9%) and CLC, 44 (71.0%). The proportion of INT was significantly correlated with gender (female-dominant) (P < 0.05), tumour size (P < 0.05), histological grading of HCC (P < 0.01) and inversely correlated with the degree of stromal fibrosis (P < 0.05). The proportion of CLC was significantly correlated with the degree of fibrosis (P < 0.01) and inflammation (P < 0.01), and inversely correlated with tumour size (P < 0.01) and histological grading of HCC (P < 0.05). The proportion of TS was significantly inversely correlated with the degree of inflammation (P < 0.01). Histological diversity score was significantly correlated with vascular invasion and the positivity for α-foetoprotein. The proportion of each SC subtype was significantly associated with certain clinicopathological factors, suggesting different properties of each SC subtypes.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, SAJ first grade, 9.5-10.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 3% in ethanol
Sigma-Aldrich
Hydrochloric acid, suitable for arsenic determination, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ super special grade, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.1 M
Sigma-Aldrich
Hydrochloric acid, suitable for determination of toxic metals, ≥35.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M