  • Search Results
  • Synthesis and anticancer activities of a novel class of mono- and di-metallic Pt(II)(salicylaldiminato)(DMSO or Picolino)Cl complexes.
Synthesis and anticancer activities of a novel class of mono- and di-metallic Pt(II)(salicylaldiminato)(DMSO or Picolino)Cl complexes.
Dalton transactions (Cambridge, England : 2003) (2014-12-17)
Faiz-Ur Rahman, Amjad Ali, Rong Guo, Yun-Chang Zhang, Hui Wang, Zhan-Ting Li, Dan-Wei Zhang
ABSTRACT
A series of novel Pt(ii) complexes [cis- and trans-Pt(ii)(salicylaldimine)(DMSO)Cl (), trans-Pt(ii)(salicylaldimine)(4-picoline)Cl (), Pt(ii)(salicylaldimine)Cl (), trans- and cis/trans-Pt2(ii)(salicylaldimine)(DMSO)2Cl2 (), trans-Pt2(ii)(salicylaldimine)(4-picoline)2Cl2 () was synthesized and characterized. The structures of -cis, -trans and were determined using a single crystal X-ray analysis. This class of Pt(ii) complexes has been studied for their in vitro cytotoxicity in multiple human cancer cell lines, including breast (MCF-7), liver (HepG2), lung (A549), colon (HCT116) and cervical (Hela) cancers. -trans, and -trans showed significant cytotoxicity to these cancer cells comparable to cisplatin. A time- and dose-dependent MTT assay revealed that these complexes can suppress cell viability and cell proliferation. Mechanistically these complexes induced pro-apoptotic gene expression such as BAX, PUMA and NOXA and thereby enhanced apoptosis. Moreover, PARP cleavage in a dose-dependent manner indicated their cytotoxic effect against cancer cells. Apoptosis of cancer cells occurred through apoptotic pathways as explained by the cytometry analysis. The DNA unwinding properties of these active Pt(ii) complexes were studied by gel electrophoresis using pBR322 plasmid DNA as a target. Changes in the morphology of cancer cells were also observed upon the addition of -trans, and -trans.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Supelco
Water, for TOC analysis
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M