  • Search Results
  • HepaRG cells as human-relevant in vitro model to study the effects of inflammatory stimuli on cytochrome P450 isoenzymes.
HepaRG cells as human-relevant in vitro model to study the effects of inflammatory stimuli on cytochrome P450 isoenzymes.
Drug metabolism and disposition: the biological fate of chemicals (2014-11-06)
Katarina Rubin, Annika Janefeldt, Linda Andersson, Zsofia Berke, Ken Grime, Tommy B Andersson
ABSTRACT
The suppression of hepatic cytochrome P450 (P450) expression during inflammatory and infectious diseases and the relief of this suppression by successful disease treatment have been previously demonstrated to impact drug disposition. To address this clinically relevant phenomenon preclinically, the effect of proinflammatory cytokines on P450 isoenzymes in human hepatocytes has been examined by several researchers. In the present study we used the human hepatoma cell line (HepaRG) and cryopreserved primary human hepatocytes to investigate the effects of various inflammatory stimuli on P450 levels with the aim of further characterizing HepaRG cells as a useful surrogate for primary hepatocytes. In this study, HepaRG cells were exposed to bacterial lipopolysaccharide (LPS), interleukin-6 (IL-6), and interleukin-18 (IL-18) for 48 or 72 hours. The effects on CYP1A2, CYP2B6, and CYP3A4 mRNA and catalytic activity (phenacetin-O-deethylase, bupropion-hydroxylase, and midazolam-1'-hydroxylase) were measured. Cryopreserved pooled plateable hepatocytes were also exposed to IL-6 or IL-18 for 48 hours, and the effects on CYP1A2, CYP2B6, and CYP3A4 mRNA levels were measured. The exposure of HepaRG cells to IL-6 and LPS resulted in suppression of CYP1A2, CYP2B6, and CYP3A4 mRNA levels as well as their catalytic activities. However, no suppression of P450 activities or mRNA levels was observed after exposure to IL-18. Similar results on CYP1A2, CYP2B6, and CYP3A4 mRNA levels were observed with primary hepatocytes. The present study indicates that different proinflammatory mediators influence the expression of P450 differentially and that HepaRG cells may be used as an alternative to human hepatocytes for studies on cytokine-mediated suppression of drug-metabolizing enzymes.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%