  • Search Results
  • A validated liquid chromatographic method for the determination of polycyclic aromatic hydrocarbons in honey after homogeneous liquid-liquid extraction using hydrophilic acetonitrile and sodium chloride as mass separating agent.
A validated liquid chromatographic method for the determination of polycyclic aromatic hydrocarbons in honey after homogeneous liquid-liquid extraction using hydrophilic acetonitrile and sodium chloride as mass separating agent.
Journal of chromatography. A (2014-12-30)
Anastasia Koltsakidou, Constantinos K Zacharis, Konstantinos Fytianos
ABSTRACT
In the present report, a simple and cost-effective method for the determination of twelve US EPA priority polycyclic aromatic hydrocarbons (PAHs) in honey samples after salting-assisted liquid-liquid extraction and UHPLC with fluorescence detection is proposed. The sample treatment is based on the usage of hydrophilic acetonitrile as extraction solvent and its phase separation under high salinity conditions. Due to the high sugar content of the samples the phase separation is promoted effortlessly. Several parameters affecting the extraction efficiency and method sensitivity including the concentration of the honey samples, the type and volume of the extraction solvent, the type and quantity of the inorganic salt, extraction time and centrifugation time was systematically investigated. The method was validated in-house according to the Commission Decision 2002/657/EC guidelines. The limit of detection (LOD) of the method lay between 0.02 and 0.04ngmL(-1) (corresponding to 0.08 and 0.16ngg(-1)) which are close to the quality criteria established by European Regulation (EC) 836/2011 concerning the PAHs in foodstuffs. The mean analytical bias (expressed as relative recoveries) in all spiking levels was acceptable being in the range of 54-118% while the relative standard deviation (RSD) was lower than 19%. The proposed method has been satisfactorily applied for the analysis of the selected PAHs residues in various honey samples obtained from Greek region.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, LR, ≥99%
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, AR, ≥99.5%
Sigma-Aldrich
Acetone, LR, ≥99%
Sigma-Aldrich
Acetone, AR, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid