  • Search Results
  • Role of GAS5 noncoding RNA in mediating the effects of rapamycin and its analogues on mantle cell lymphoma cells.
Role of GAS5 noncoding RNA in mediating the effects of rapamycin and its analogues on mantle cell lymphoma cells.
Clinical lymphoma, myeloma & leukemia (2014-04-08)
Mirna Mourtada-Maarabouni, Gwyn T Williams
ABSTRACT
Inhibition of the mammalian target of rapamycin (mTOR) pathway is a promising strategy for the treatment of mantle cell lymphoma (MCL). ncRNA growth arrest-specific 5 (GAS5), a 5' terminal oligopyrimidine (5'TOP) RNA regulated by the mTOR pathway, is necessary and sufficient for normal growth arrest in leukemic and untransformed human lymphocytes. We downregulated endogenous GAS5 in mantle cell lymphoma cell lines using RNA interference before treatment with several rapalogues. The effect of GAS5 downregulation was monitored by 3 independent analyses of cell viability, DNA synthesis, and colony-forming ability. Downregulation of GAS5 substantially reduced the effects of each rapalogue on cell viability, DNA synthesis, and colony-forming ability. Stimulation of expression of candidate tumor suppressor GAS5 is responsible for much of the cytotoxic and cytostatic effects of rapalogues in MCL, suggesting that improved targeting of this pathway may allow improvements in the therapy of this intractable lymphoma.
Product Description
Supelco
Rapamycin, VETRANAL®, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, JIS special grade, ≥99.0%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus