  • Search Results
  • An inhalation anesthetic device for stereotaxic operation on mouse pups.
An inhalation anesthetic device for stereotaxic operation on mouse pups.
Journal of neuroscience methods (2015-02-04)
Sachine Yoshida, Yuya Morimoto, Taishi Tonooka, Shoji Takeuchi
ABSTRACT
Mouse pups are invaluable model animals for understanding the molecular and neural basis underlying behavioral development. Stereotaxic operations with anesthetic control are useful tools in systems neuroscience. However, there are no commercially available anesthetic or stereotaxic devices for mouse pups. Current devices have several problems such as invasive approach for stabilization, poor sanitary control, and less flexibility to combine other surgical apparatuses. Here, we developed an inhalation anesthetic device equipped with stereotaxic function for mouse pups, by using polydimethylsiloxane (PDMS). PDMS is tolerant to heat and water exposure, and soft enough to cut or make a hole. The anesthetic and the stereotaxic parts were fabricated from the three-dimensional computer-aided design (3D CAD) data obtained from the head of a real mouse pup. To confirm its utility, a tracer was injected into the brain. We were able to anesthetize and stabilize pups at once in a non-invasive manner using the PDMS device. The histological staining revealed that tracer injection was successful. Our device was compatible with various types of commercial stereotaxic and anesthetic apparatuses via trimming and tube insertion, respectively. To our knowledge, this is the first report of a device that can stabilize the mouse pup's head with the non-invasive manner and functions as an inhalation anesthetic device that can be sterilized. The present fabrication method will provide a handy and functional instrument for stereotaxic operations in animal models at various developmental stages.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, JIS 1000, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ≥99.8%, suitable for absorption spectrum analysis
Sigma-Aldrich
Methanol, ≥99.8%, for chromatography
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Supelco
Methanol, analytical standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%