Merck
  • Preclinical validation of AXL receptor as a target for antibody-based pancreatic cancer immunotherapy.
Preclinical validation of AXL receptor as a target for antibody-based pancreatic cancer immunotherapy.
Oncogene (2013-11-19)
W Leconet, C Larbouret, T Chardès, G Thomas, M Neiveyans, M Busson, M Jarlier, N Radosevic-Robin, M Pugnière, F Bernex, F Penault-Llorca, J-M Pasquet, A Pèlegrin, B Robert
ABSTRACT
AXL receptor tyrosine kinase (RTK) is implicated in proliferation and invasion of many cancers, particularly in pancreatic ductal adenocarcinoma (PDAC), for which new therapeutic options are urgently required. We investigated whether inhibition of AXL activity by specific monoclonal antibodies (mAbs) is efficient in limiting proliferation and migration of pancreatic cancer cells. Expression of AXL was evaluated by immunohistochemistry in 42 PDAC. The AXL role in oncogenesis was studied using the short hairpin RNA approach in a pancreatic carcinoma cell line. We further generated antihuman AXL mAbs and evaluated their inhibitory effects and the AXL downstream signaling pathways first in vitro, in a panel of pancreatic cancer cell lines and then in vivo, using subcutaneous or orthotopic pancreatic tumor xenografts. AXL receptor was found expressed in 76% (32/42) of PDAC and was predominantly present in invasive cells. The AXL-knockdown Panc-1 cells decreased in vitro cell migration, survival and proliferation, and reduced in vivo tumor growth. Two selected anti-AXL mAbs (D9 and E8), which inhibited phosphorylation of AXL and of its downstream target AKT without affecting growth arrest-specific factor 6 (GAS6) binding, induced downexpression of AXL by internalization, leading to an inhibition of proliferation and migration in the four pancreatic cancer cell lines studied. In vivo, treatment by anti-AXL mAbs significantly reduced growth of both subcutaneous and orthotopic pancreatic tumor xenografts independently of their KRAS mutation status. Our in vitro and preclinical in vivo data demonstrate that anti-human AXL mAbs could represent a new approach to the pancreatic cancer immunotherapy.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
DAPI, for nucleic acid staining
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%