  • Search Results
  • Dose-dependent effects of the antiprogestin, RU486, on sexual behavior of naturally cycling Fischer rats.
Dose-dependent effects of the antiprogestin, RU486, on sexual behavior of naturally cycling Fischer rats.
Behavioural brain research (2015-01-17)
Lynda Uphouse
ABSTRACT
Regularly cycling Fischer female rats were treated with either a low (5mg/kg) or high (5mg/RAT; approximately 30mg/kg) dose of the antiprogestin, RU486, before the morning of proestrus or on the morning of proestrus. The emergence of sexual behavior after treatment with RU486 was examined in a mating test with a sexually active male rat. Lordosis behavior was remarkably resistant to the effects of RU486. Only the high dose of RU486 given the evening before proestrus, approximately 22h before mating, reduced lordosis behavior. Independent of dose or time of treatment, proceptivity was reduced and resistance to the male's attempts to mount was increased by RU486 treatment. In addition, the effect of a 5min restraint stress on sexual behavior was examined. In contrast to the relative resistance of lordosis behavior of unrestrained rats to RU486 treatment, RU486 treated rats showed a significant decline in lordosis behavior after restraint. These findings allow the suggestion that the emergence of lordosis behavior is relatively resistant to the antiprogestin while the maintenance of lordosis behavior after restraint may require participation of intracellular progesterone receptors.
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
1,2-Propanediol, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard