Merck
  • How safe is acetaminophen use in patients treated with vitamin K antagonists? A systematic review and meta-analysis.
How safe is acetaminophen use in patients treated with vitamin K antagonists? A systematic review and meta-analysis.
Thrombosis research (2014-12-03)
Daniel Caldeira, João Costa, Márcio Barra, Fausto J Pinto, Joaquim J Ferreira
ABSTRACT
Acetaminophen is a commonly prescribed and over-the-count used drug, and is considered to be the preferred treatment choice for anticoagulated patients requiring analgesic drug therapy. However, observational data have suggested that this drug combination may increase the International Normalized Ratio (INR) values and bleeding events in patients taking Vitamin K antagonists (VKAs). Still, the clinical impact of this putative effect remains unknown. Therefore, we performed a systematic review of randomized controlled trials (RCTs) to estimate the impact of concomitant use of acetaminophen and VKA in the INR measurements Systematic review and meta-analysis of RCTs comparing acetaminophen versus placebo or no treatment, in VKA-treated patients and reporting INR estimates. Medline and Cochrane Library were searched up to April 2014. Primary outcome was the mean difference (MD) between the greatest INR elevations in each treatment arm. Random-effects meta-analysis was performed to derive pooled estimates and 95% Confidence Interval (CI). Heterogeneity was evaluated with I(2) test. Seven RCTs (n=225 patients) were included. Acetaminophen was associated with a mean 0.62 INR increase (95%CI: 0.46 to 0.78; I(2)=25%) compared to placebo in VKA-treated patients. Studies did not report any major bleeding event. Meta-regression showed a significant 0.17 mean increase of the INR per each daily gram of acetaminophen (95%CI: 0.004 to 0.33). Acetaminophen is associated with a statistically significant and possible clinically relevant increase in the INR, with a dose dependent relationship. Patients treated concomitantly with VKA and acetaminophen should be monitored more regularly for possible VKA dosage adjustment.
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®