  • Search Results
  • Development and characterization of a solid dispersion film for the vaginal application of the anti-HIV microbicide UAMC01398.
Development and characterization of a solid dispersion film for the vaginal application of the anti-HIV microbicide UAMC01398.
International journal of pharmaceutics (2014-09-02)
Carolien Grammen, Guy Van den Mooter, Bernard Appeltans, Johan Michiels, Tania Crucitti, Kevin K Ariën, Koen Augustyns, Patrick Augustijns, Joachim Brouwers
ABSTRACT
The purpose of this work was to design and evaluate a vaginal film delivery system for UAMC01398, a novel non-nucleoside reverse transcriptase inhibitor currently under investigation for use as an anti-HIV microbicide. UAMC01398 (1mg) films consisting of hydroxypropylmethylcellulose (HPMC) and polyethylene glycol 400 (PEG400) in different ratios were prepared by solvent evaporation. Based on its flexibility, softness and translucent appearance, the 30% PEG400 and 70% HPMC containing film was selected for further assessment. The vaginal film formulation was fast-dissolving (<10 min in 1 mL of vaginal fluid simulant), stable up to at least one month and safe toward epithelial cells and lactobacilli. Furthermore, formulating UAMC01398 into the film dosage form did not influence its antiviral activity. Powder X-ray diffraction revealed the amorphous nature of the UAMC01398 film, resulting in enhanced compound permeation across the epithelial HEC-1A cell layer, presumably owing to the induction of supersaturation. The in vivo vaginal tissue uptake of UAMC01398 in rabbits, as measured by systemic concentrations, was increased compared to the previously established non-solubilizing gel (significant difference) and sulfobutyl ether-β-cyclodextrin (5%) containing gel. To conclude, we identified a film formulation suitable for the vaginal delivery of UAMC01398.
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), suitable for iron determination