  • Search Results
  • Autophagy is induced by raptor degradation via the ubiquitin/proteasome system in granular corneal dystrophy type 2.
Autophagy is induced by raptor degradation via the ubiquitin/proteasome system in granular corneal dystrophy type 2.
Biochemical and biophysical research communications (2014-07-22)
Seung-Il Choi, Yong-Sun Maeng, Kyu Seo Kim, Tae-Im Kim, Eung Kweon Kim
ABSTRACT
Granular corneal dystrophy type 2 (GCD2) is an autosomal dominant disorder that is caused by a point mutation in transforming growth factor-β-induced gene-h3 (TGFBI), which encodes transforming growth factor-β-induced protein (TGFBIp). Recently, we found that the autophagic clearance of mutant-TGFBIp is delayed in GCD2 corneal fibroblasts; however, any potential correlation between mutant-TGFBIp turnover and autophagy-lysosome pathway remains unknown. Here, we report that mutant-TGFBIp is accumulated and that autophagy, a key clearance pathway for mutant-TGFBIp, is induced in primary cultured GCD2 homozygous (HO) and wild-type (WT) corneal fibroblasts that express exogenously introduced mutant-TGFBIp. Mutant-TGFBI colocalized with LC3-enriched cytosolic vesicles and cathepsin D in primary cultured GCD2 corneal fibroblasts. We also observed reduced levels of raptor (regulatory-associated protein of the mammalian target of rapamycin [mTOR]) in GCD2 corneal fibroblasts and WT corneal fibroblasts expressing mutant-TGFBIp. Strikingly, treatment with MG132, a ubiquitin/proteasome system inhibitor, significantly increased the levels of both total and ubiquitinated raptor in GCD2 corneal fibroblasts. The levels of the autophagy marker LC3-II were also increased in WT corneal fibroblasts that were treated with shRNA against raptor. However, mutant-TGFBIp accumulated in autophagosomes or/and lysosomes in spite of the significant activation of basal autophagy in GCD2 corneal fibroblasts. These results suggest that an insufficient autophagy-lysosome pathway might be responsible for the intracellular accumulation of mutant-TGFBIp during the pathogenesis of GCD2.
Product Description
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Dimethyl sulfoxide, analytical standard
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)