Merck
  • Development of an alternative non-obese non-genetic rat model of type 2 diabetes using caffeine and streptozotocin.
Development of an alternative non-obese non-genetic rat model of type 2 diabetes using caffeine and streptozotocin.
Pharmacological reports : PR (2014-06-21)
Pragalathan Naidoo, Md Shahidul Islam
ABSTRACT
The aim of the present study was to develop an alternative non-obese non-genetic rat model of type 2 diabetes (T2D). Six-week-old male SD rats were randomly divided into six groups, namely: Normal Control (NC), Diabetic Control (DBC), Caffeine 5mg/kg BW+STZ (CAF5), Caffeine 10mg/kg BW+STZ (CAF10), Caffeine 20mg/kg BW+STZ (CAF20) and Caffeine 40mg/kg BW+STZ (CAF40) and were fed a normal rat pellet diet and drinking water ad libitum throughout the experimental period. After a one week acclimatization period, diabetes was induced in the animals in DBC and all CAF groups with an injection (i.p.) of the respective dosages of caffeine (mg/kg BW) 15min before the injection of STZ (65mg/kg BW) when normal saline was injected to the DBC group instead of caffeine. The NC group received normal saline and buffer instead of caffeine and STZ, respectively. One week after the STZ injection, animals with non-fasting blood glucose>300mg/dl were considered as diabetic. Three weeks after the STZ injection, the animals in the CAF5 and CAF10 groups were eliminated from the study due to the severity of diabetes and the experiment was continued with the remainder groups for a 13 weeks period. The data of food and fluid intake, body weight, blood glucose, glucose tolerance test, HOMA-IR, HOMA-beta, serum insulin, fructosamine, lipid profile and organ specific enzymes, anti-diabetic drug response tests, and pancreatic histopathology suggest that CAF20 group can be a better alternative non-genetic model of non-obese T2D.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®