  • Search Results
  • Comparative bioavailability of rifampicin and isoniazid in fixed-dose combinations and single-drug formulations.
Comparative bioavailability of rifampicin and isoniazid in fixed-dose combinations and single-drug formulations.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease (2014-12-18)
L-H Hao, S-C Guo, C-C Liu, H Zhu, B Wang, L Fu, M-T Chen, L Zhou, J-Y Chi, W Yang, W-J Nie, Y Lu
ABSTRACT
The bioavailability of rifampicin (RMP) decreases by ∼30% on interaction with isoniazid (INH) in stomach acid conditions, which can result in the development of drug resistance and treatment failure. To compare the bioavailability in healthy volunteers of five anti-tuberculosis fixed-drug combinations (FDCs) used in China (formulations A-E) containing RMP and INH against single-drug formulations taken as reference. Two- or three-period, two- or three-sequence crossover study of drugs. Only RMP formulation E passed the bioequivalence criteria, with 90% confidence intervals for the log-transformed ratios of AUC₀₋₂₄, AUC₀₋∞, and Cmax of respectively 89.9-103.7, 89.6-102.2 and 87.7-107.9. For INH, formulations A, B, C and D passed the bioequivalence test, but not product E, where the 90%CIs of the log-transformed ratios of AUC₀₋₂₄, AUC₀₋∞, and Cmax were respectively 85.2-100.7, 85.2-100.7 and 73.8-100.9. According to the results of the bioequivalence analysis carried out in this study, RMP formulations A, B, C and D were not within the acceptable range and only formulation E passed the bioequivalence criteria of 80-125%. In comparison, four-test INH formulations (A, B, C and D) were bioequivalent to the corresponding single-drug formulation, while product E failed in the bioequivalence criteria.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Acetaminophen, analytical standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard