  • Search Results
  • CKIP-1 acts as a colonic tumor suppressor by repressing oncogenic Smurf1 synthesis and promoting Smurf1 autodegradation.
CKIP-1 acts as a colonic tumor suppressor by repressing oncogenic Smurf1 synthesis and promoting Smurf1 autodegradation.
Oncogene (2013-09-03)
J Nie, L Liu, G Xing, M Zhang, R Wei, M Guo, X Li, P Xie, L Li, F He, W Han, L Zhang
ABSTRACT
Dysregulation of cellular signaling pathways can lead to colon cancer. However, research on the key signaling effectors or regulators in colon carcinogenesis is limited. Casein kinase-2 interacting protein-1 (CKIP-1; also known as PLEKHO1) is crucial during adult bone formation and is a promising drug target for osteoporosis therapy. In this study, we observed that CKIP-1 was downregulated in human colon cancer tissues and colon cancer cell lines, and this result was correlated with colon cancer progression. CKIP-1 silencing in colon cancers involved promoter methylation. In colon cancer HCT116 and SW480 cells, CKIP-1 overexpression inhibited cell growth and migration. CKIP-1 also suppressed in-vivo tumor formation. Notably, the growth-suppressive role of CKIP-1 was dependent on the downregulation of the cell cycle-regulated oncogene Smad ubiquitylation regulatory factor-1 (Smurf1). During cell cycle progression, phosphatidylinositol-3-kinase (PI3K)/Akt/mammalian target of rapamycin (mTOR) signaling increased Smurf1 production by an mTOR-dependent translational control mechanism. Rapamycin, the mTOR inhibitor, significantly reduced Smurf1 protein levels, and Smurf1 was degraded in mitosis. In colon cancer, CKIP-1 controlled Smurf1 expression by suppressing PI3K/Akt/mTOR signaling and enhancing Smurf1 autodegradation, and CKIP-1 downregulation was correlated with Smurf1 upregulation in colon carcinogenesis. These findings provide novel insight into the mechanisms of the candidate tumor suppressor CKIP-1.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)